Tisk…

8-Hydroxyquinoline-β-D-glucuronic acid, sodium salt

Dodavatel: Apollo Scientific
Varování

BIB6070-1G BIB6070-5G BIB6070-500MG
APOSBIB6070-1GEA 13560 CZK
APOSBIB6070-1G APOSBIB6070-5G APOSBIB6070-500MG
8-Hydroxyquinoline-β-D-glucuronic acid, sodium salt
8-Hydroxyquinoline-β-D-glucuronic acid, sodium salt
Vzorec: C₁₅H₁₄NO₇·Na
MW: 343.264 g/mol
Storage Temperature: Freezer
MDL Number: MFCD02683911
CAS číslo: 207728-71-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR