Tisk…

5-Bromo-3-indolyl phosphate, p-toluidine salt

Dodavatel: Apollo Scientific
Varování

BIB6014-1G BIB6014-500MG BIB6014-5G
APOSBIB6014-1GEA 2480 CZK
APOSBIB6014-1G APOSBIB6014-500MG APOSBIB6014-5G
5-Bromo-3-indolyl phosphate, p-toluidine salt
5-Bromo-3-indolyl phosphate, p-toluidine salt
Vzorec: C₈H₇BrNO₄P·C₇H₉N
Storage Temperature: Freezer
MDL Number: MFCD03427361
CAS číslo: 80008-69-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR