5-Bromo-3-indolyl caprylate
Dodavatel: Apollo Scientific
|
Varování
|
BIB6011-500MG
APOSBIB6011-500MGEA
4140
CZK
APOSBIB6011-500MG
5-Bromo-3-indolyl caprylate
5-Bromo-3-indolyl caprylate
Vzorec:
C₁₆H₂₀BrNO₂ MW: 338,239 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD02683904 CAS číslo: 133950-69-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...