Tisk…

5-Bromo-4-chloro-3-indolyl N-acetyl-β-D-galactosaminide

Dodavatel: Apollo Scientific
Varování

Synonyms: X-GalNAc

BIB1741-100MG BIB1741-1G BIB1741-250MG
APOSBIB1741-100MGEA 2980 CZK
APOSBIB1741-100MG APOSBIB1741-1G APOSBIB1741-250MG
5-Bromo-4-chloro-3-indolyl N-acetyl-β-D-galactosaminide
5-Bromo-4-chloro-3-indolyl N-acetyl-β-D-galactosaminide
Vzorec: C₁₆H₁₈BrClN₂O₆
MW: 449,681 g/mol
Bod tání: 220 °C
Storage Temperature: Freezer
MDL Number: MFCD00083260
CAS číslo: 129572-48-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR