Tisk…

L-Lysine 7-amido-4-methylcoumarin acetate salt

Dodavatel: Apollo Scientific
Varování

Synonyms: L-Lysine-AMC acetate salt

BIB1442-100MG BIB1442-250MG BIB1442-50MG
APOSBIB1442-100MGEA 3310 CZK
APOSBIB1442-100MG APOSBIB1442-250MG APOSBIB1442-50MG
L-Lysine 7-amido-4-methylcoumarin acetate salt
L-Lysine 7-amido-4-methylcoumarin acetate salt
Vzorec: C₁₆H₂₁N₃O₃·CH₃CO₂H
MW: 363,408 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00152123
CAS číslo: 201853-23-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR