1-Naphthyl N-acetyl-β-D-glucosaminide
Dodavatel: Apollo Scientific
|
Varování
|
BIB1425-250MG
BIB1425-500MG
APOSBIB1425-250MGEA
4370
CZK
APOSBIB1425-250MG
APOSBIB1425-500MG
1-Naphthyl N-acetyl-β-D-glucosaminide
1-Naphthyl N-acetyl-β-D-glucosaminide
Vzorec:
C₁₈H₂₁NO₆ MW: 347.362 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00051151 CAS číslo: 10329-98-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...