Tisk…

6-Fluoro-3-indolyl β-D-galactopyranoside

Dodavatel: Apollo Scientific
Varování

BIB1420-25MG BIB1420-50MG BIB1420-100MG
APOSBIB1420-25MGEA 10750 CZK
APOSBIB1420-25MG APOSBIB1420-50MG APOSBIB1420-100MG
6-Fluoro-3-indolyl β-D-galactopyranoside
6-Fluoro-3-indolyl β-D-galactopyranoside
Vzorec: C₁₄H₁₆FNO₆
MW: 313,278 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00798396
CAS číslo: 207727-11-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR