5-Bromo-6-chloro-3-indolyl α-D-galactopyranoside
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Magenta-α-gal
BIB1416-1G
BIB1416-250MG
BIB1416-2G
APOSBIB1416-1GEA
6220
CZK
APOSBIB1416-1G
APOSBIB1416-250MG
APOSBIB1416-2G
5-Bromo-6-chloro-3-indolyl α-D-galactopyranoside
5-Bromo-6-chloro-3-indolyl α-D-galactopyranoside
Vzorec:
C₁₄H₁₅BrClNO₆ MW: 408,629 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD02683897 CAS číslo: 198402-60-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...