Tisk…

5-Bromo-6-chloro-3-indolyl N-acetyl-β-D-glucosaminide

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Bromo-6-chloro-3-indoxyl-2-acetamido-2-deoxy-β-D-glucopyranoside, Magenta N-acetyl-β-D-glucosaminide

BIB1415-100MG BIB1415-250MG
APOSBIB1415-100MGEA 1870 CZK
APOSBIB1415-100MG APOSBIB1415-250MG
5-Bromo-6-chloro-3-indolyl N-acetyl-β-D-glucosaminide
5-Bromo-6-chloro-3-indolyl N-acetyl-β-D-glucosaminide
Vzorec: C₁₆H₁₈BrClN₂O₆
MW: 449,681 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00798384
CAS číslo: 5609-91-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR