6-Chloro-3-indolyl α-D-mannopyranoside
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Salmon-α-mannoside
BIB1414-100MG
BIB1414-250MG
BIB1414-25MG
APOSBIB1414-100MGEA
7610
CZK
APOSBIB1414-100MG
APOSBIB1414-250MG
APOSBIB1414-25MG
6-Chloro-3-indolyl α-D-mannopyranoside
6-Chloro-3-indolyl α-D-mannopyranoside
Vzorec:
C₁₄H₁₆ClNO₆ MW: 329,733 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD02683909 CAS číslo: 425427-88-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...