6-Chloro-3-indolyl β-D-glucopyranoside
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Salmon-glucoside
BIB1412-1G
BIB1412-500MG
BIB1412-5G
APOSBIB1412-1GEA
3740
CZK
APOSBIB1412-1G
APOSBIB1412-500MG
APOSBIB1412-5G
6-Chloro-3-indolyl β-D-glucopyranoside
6-Chloro-3-indolyl β-D-glucopyranoside
Vzorec:
C₁₄H₁₆ClNO₆ MW: 329,733 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00467206 CAS číslo: 159954-28-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...