Tisk…

6-Chloro-3-indolyl N-acetyl-β-D-glucosaminide

Dodavatel: Apollo Scientific
Varování

Synonyms: Salmon-N-acetyl-β-D-glucosaminide

BIB1410-25MG BIB1410-50MG
APOSBIB1410-25MGEA 1370 CZK
APOSBIB1410-25MG APOSBIB1410-50MG
6-Chloro-3-indolyl N-acetyl-β-D-glucosaminide
6-Chloro-3-indolyl N-acetyl-β-D-glucosaminide
Vzorec: C₁₆H₁₉ClN₂O₆
Storage Temperature: Freezer
MDL Number: MFCD04972051
CAS číslo: 156117-44-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR