Tisk…

5-Bromo-4-chloro-3-indolyl caprylate

Dodavatel: Apollo Scientific
Varování

Synonyms: X-caprylate

BIB1401-250MG BIB1401-1G BIB1401-5G
APOSBIB1401-250MGEA 2650 CZK
APOSBIB1401-250MG APOSBIB1401-1G APOSBIB1401-5G
5-Bromo-4-chloro-3-indolyl caprylate
5-Bromo-4-chloro-3-indolyl caprylate
Vzorec: C₁₆H₁₉BrClNO₂
MW: 372,685 g/mol
Bod tání: 57…60 °C
Storage Temperature: Freezer
MDL Number: MFCD00083263
CAS číslo: 129541-42-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR