Tisk…

Arachidoyl-DL-carnitine chloride

Dodavatel: Apollo Scientific
Varování

BIA4018-250MG BIA4018-100MG
APOSBIA4018-250MGEA 21100 CZK
APOSBIA4018-250MG APOSBIA4018-100MG
Arachidoyl-DL-carnitine chloride
Arachidoyl-DL-carnitine chloride
Vzorec: C₂₇H₄₆NO₄·Cl
MW: 484,111 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00274384
CAS číslo: 149116-07-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR