Aflunox™ 2507
Dodavatel: Apollo Scientific
|
Varování
|
PC1002N-100G
PC1002N-25G
APOSPC1002N-100GEA
1780
CZK
APOSPC1002N-100G
APOSPC1002N-25G
Aflunox™ 2507
Aflunox™
Perfluoropolyether fluids are inert to aggressive reagents including elemental fluorine. Aflunox 2507 is an excellent inert lubricant for laboratory stirrer seals.
- F[CF(CF3)CF2O]xC2F5 x = 21
- viscosity 270 cSt@20C
Aflunox is a registered trademark of Japan Mektron
Bod tání:
-45 °C Hustota: 1,89 g/cm3 @ 25°C |
MDL Number:
MFCD00166448 CAS číslo: 52700-35-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...