Aflunox™ 606 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC1002E-100G
PC1002E-25G
APOSPC1002E-100GEA
1620
CZK
APOSPC1002E-100G
APOSPC1002E-25G
Aflunox™ 606 95%
Aflunox™
Aflunox oils are clear colourless oils suitable for use as lubricants and are inert to oxidising agents such as fluorine, chlorine and oxygen at high temperatures.
- F[CF(CF3)CF2O]xC2F5 x = 12
- viscosity 68 cst@20C
Aflunox is a registered trademark of Japan Mektron
Bod tání:
–60 °C Hustota: 1,88 g/cm3 @ 25°C |
MDL Number:
MFCD00166448 CAS číslo: 52700-35-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...