Tisk…

Polyethylenglykol 1500, GPR RECTAPUR®

Dodavatel: VWR Chemicals

Synonyms: PEG 1500, PEG 1,500

26607.290EA 1140 CZK
26607.290 26607.360
Polyethylenglykol 1500, GPR RECTAPUR®
Polyethylenglykol 1500
Vzorec: H(OCH₂CH₂)nOH
MW: 1500 g/mol
MDL Number: MFCD00081839
CAS číslo: 25322-68-3

Objednat

Výsledky specifikačního testu

Average molecular weight 1400 - 1600
Hydroxyl value 70 - 80
pH (5%) 4,0 - 8,0
Viscosity (20°C, 50% water) 36 - 42 mPa.s
Solidification point about 45°C
Colouration (25% water) max 50 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR