Tisk…
Paladium 5% on carbon Type 87L wetted with ca. 50% water
Dodavatel: Thermo Fisher Scientific
Varování
H33434.09 H33434.18
H33434.09EA 5670 CZK
H33434.09 H33434.18
Paladium 5% on carbon Type 87L wetted with ca. 50% water
Palladium
Vzorec: Pd
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS číslo: 7440-05-3