EH-IDTBr, Organic Photovoltaics (OPV) reagent
Dodavatel: Thermo Scientific
Synonyms:
5,5'-(((4,4,9,9-Tetrakis(2-ethylhexyl)-4,9-dihydro-s-indaceno[1,2-b:5,6-b']dithiophene-2,7-diyl)bis(benzo[c][1,2,5]thiadiazole-7,4-diyl))bis(methaneylylidene))bis(3-ethyl-2-thioxothiazolidin-4-one)
H66142.MC
H66142.MD
H66142.ME
H66142.MCEA
23100
CZK
H66142.MC
H66142.MD
H66142.ME
EH-IDTBr, Organic Photovoltaics (OPV) reagent
EH-IDTBr
Vzorec:
C₇₂H₈₈N₆O₂S₈ MW: 1326,03 g/mol |
CAS číslo:
2102510-60-9 |
Výsledky specifikačního testu
Appearance (Color) | Black |
Form | Flake or threadlike |
Bandgap energy | 1,64 ± 0,02 eV |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...