Tisk…

PDCBT, Organic Photovoltaics (OPV) reagent

Dodavatel: Thermo Scientific

Synonyms: Poly[5,5'-bis(2-butyloctyl)-(2,2'-bithiophene)-4,4'-dicarboxylate-alt-5,5'-2,2'- bithiophene]

H66713.MC H66713.MD H66713.ME
H66713.MCEA 23100 CZK
H66713.MC H66713.MD H66713.ME
PDCBT, Organic Photovoltaics (OPV) reagent
PDCBT
Vzorec: (C₄₄H₆₂O₄S₄)n
CAS číslo: 1609536-17-5

Objednat

Výsledky specifikačního testu

Appearance (Color) Black
Form Flake or threadlike
Bandgap energy 1,89 ± 0,02 eV

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR