Eriodictyol
Dodavatel: Thermo Fisher Scientific
|
Varování
|
465240010
465242500
ACRO465240010EA
19750
CZK
ACRO465240010
ACRO465242500
Eriodictyol
Eriodictyol
- Bioactive
Vzorec:
C₁₅H₁₂O₆ Storage Temperature: Refrigerator |
MDL Number:
MFCD00135890 CAS číslo: 552-58-9 |
Výsledky specifikačního testu
Appearance | White to light beige to light brown Powder or crystals |
Infrared spectrum | Conforms |
Water | ≤5 % |
HPLC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...