Sitafloxacin sesquihydrate
Dodavatel: Thermo Scientific
|
Varování
|
464370010
464372500
ACRO464370010EA
11510
CZK
ACRO464370010
ACRO464372500
Sitafloxacin sesquihydrate
Sitafloxacin sesquihydrate
- Bioactive
Vzorec:
C₁₉H₁₈ClF₂N₃O₃·1.5H₂O MW: 436,84 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00903720 CAS číslo: 163253-35-8 |
Výsledky specifikačního testu
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | 5.5 % to 8.0 % |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...