Divalproex sodium
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Valproic acid (sodium)(2:1), Sodium hydrogen bis(2-propylpentanoate)
463490250
463491000
ACRO463490250EA
3490
CZK
ACRO463490250
ACRO463491000
Divalproex sodium
Divalproex sodium
- Bioactive
Vzorec:
C₁₆H₃₁NaO₄ MW: 310.4 g/mol Bod tání: 98…102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00879852 CAS číslo: 76584-70-8 |
Výsledky specifikačního testu
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 98 to 102 °C |
Water | ≤1 % |
HPLC | ≥97.5 % |
Sodium (Na) | passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...