Tisk…

Ibandronate sodium salt monohydrate

Dodavatel: Thermo Fisher Scientific

463150010 463150050
ACRO463150010EA 2020 CZK
ACRO463150010 ACRO463150050
Ibandronate sodium salt monohydrate
Ibandronate sodium salt monohydrate

  • Bioactive

Vzorec: C₉H₂₂NNaO₇P₂·H₂O
MW: 359.23 g/mol
Bod tání: 175…177 °C
Storage Temperature: Freezer
MDL Number: MFCD07197214
CAS číslo: 138926-19-9

Objednat

Výsledky specifikačního testu

Appearance White to almost white Powder
Infrared spectrum Conforms
Loss on drying 4.5 % to 8.5 %
Titration with NaOH 96 to 104 % (on dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR