Tisk…

Cidofovir dihydrate

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: (S)-(((1-(4-Amino-2-oxopyrimidin-1(2H)-yl)-3-hydroxypropan-2-yl)oxy)methyl)phosphonic acid dihydrate

463120010 463122500
ACRO463120010EA 25120 CZK
ACRO463120010 ACRO463122500
Cidofovir dihydrate
Cidofovir dihydrate

  • Bioactive

Vzorec: C₈H₁₄N₃O₆P·2H₂O
MW: 315,22 g/mol
Bod tání: 260 °C
Storage Temperature: Freezer
MDL Number: MFCD16556167
CAS číslo: 149394-66-1
UN: 2811
ADR: 6.1,III

Objednat

Výsledky specifikačního testu

Appearance White to almost white Powder
Infrared spectrum Conforms
Water 10 to 12.5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR