Tisk…

Tylosin phosphate

Dodavatel: Thermo Fisher Scientific
Nebezpečí

463040010 463040050
ACRO463040010EA 2980 CZK
ACRO463040010 ACRO463040050
Tylosin phosphate
Tylosin phosphate

  • Bioactive

Vzorec: C₄₆H₇₇NO₁₇·H₃PO₄
MW: 1014.1 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06795692
CAS číslo: 1405-53-4

Objednat

Výsledky specifikačního testu

Appearance White to light yellow Powder
Infrared spectrum Conforms
Solubility (1% in water) Clear colorless to light yellow
HPLC ≥94 % (sum of tylosins A+B+C+D)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR