Tisk…

Chromazurol B (synthetic)

Dodavatel: Thermo Fisher Scientific

Synonyms: Mordant Blue 1, Eriochrome Azurol B, Sunchromine Pure Blue BX

464530050 464530250
ACRO464530050EA 1270 CZK
ACRO464530050 ACRO464530250
Chromazurol B (synthetic)
Chromazurol B

  • Bioactive

Vzorec: C₂₃H₁₄Cl₂Na₂O₆
Storage Temperature: Ambient
MDL Number: MFCD00036446
CAS číslo: 1796-92-5
EINECS: 217-279-1

Objednat

Výsledky specifikačního testu

Appearance Orange-red to brown Powder
Infrared spectrum Conforms
Absorptivity (1%, 1cm) ≥220
UV Lambda max 423 to 429 nm (1% water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR