Tisk…

Carteolol hydrochloride

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 5-[3-[(1,1-Dimethylethyl)amino]-2-hydroxypropoxy]-3,4-dihydro-2(1H)-quinolinone hydrochloride

464320010 464320050
ACRO464320010EA 4010 CZK
ACRO464320010 ACRO464320050
Carteolol hydrochloride
Carteolol hydrochloride

  • Bioactive

Vzorec: C₁₆H₂₄N₂O₃·HCl
Bod tání: 278 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00941499
CAS číslo: 51781-21-6

Objednat

Výsledky specifikačního testu

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (1 g, 105°C)
Solubility (0.3 g in 10 ml water) Clear colorless solution
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR