Tisk…

Nobiletin

Dodavatel: Thermo Fisher Scientific

463520010 463522500
ACRO463520010EA 12650 CZK
ACRO463520010 ACRO463522500
Nobiletin
Nobiletin

  • Bioactive

Vzorec: C₂₁H₂₂O₈
Bod tání: 138 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03273560
CAS číslo: 478-01-3

Objednat

Výsledky specifikačního testu

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 136 to 140 °C
Water ≤5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR