Tisk…

(6R,7R)-7-((Z)-2-(2-Aminothiazol-4-yl)-2-(methoxyimino)acetamido)-3-(((furan-2-carbonyl)thio)methyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrochloride

Dodavatel: Thermo Fisher Scientific

463420010 463420050 463420250
ACRO463420010EA 1280 CZK
ACRO463420010 ACRO463420050 ACRO463420250
(6R,7R)-7-((Z)-2-(2-Aminothiazol-4-yl)-2-(methoxyimino)acetamido)-3-(((furan-2-carbonyl)thio)methyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrochloride
(6R,7R)-7-((Z)-2-(2-Aminothiazol-4-yl)-2-(methoxyimino)acetamido)-3-(((furan-2-carbonyl)thio)methyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrochloride

  • Bioactive

Vzorec: C₁₉H₁₈ClN₅O₇S₃
Bod tání: >190 °C
Storage Temperature: Refrigerator
CAS číslo: 103980-44-5

Objednat

Výsledky specifikačního testu

Appearance White to light yellow to light beige Powder or crystals
Infrared spectrum Conforms
Water ≤5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR