Tisk…

Repaglinide

Dodavatel: Thermo Scientific

Synonyms: 2-Ethoxy-4-[2-[[(1S)-3-methyl-1-[2-(1-piperidinyl)phenyl]butyl]amino]-2-oxoethyl]benzoic acid

463230010 463232500
ACRO463230010EA 12390 CZK
ACRO463230010 ACRO463232500
Repaglinide
Repaglinide

  • Bioactive

Vzorec: C₂₇H₃₆N₂O₄
MW: 452,59 g/mol
Bod tání: 129…130 °C
Storage Temperature: Freezer
MDL Number: MFCD00906179
CAS číslo: 135062-02-1

Objednat

Výsledky specifikačního testu

Appearance White to almost white Powder
Infrared spectrum Conforms
Melting point 129 to 135 °C
Loss on drying ≤0.5 % (105 °C)
HPLC ≥97.5 %
Specific optical rotation +7° to +10° (20 °C, 589 nm) (1% in MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR