Tisk…

(+)-Catechin monohydrate

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: (+)-3,3’,4’,5,7-Flavanpentol.H₂O

463020010 463020050
ACRO463020010EA 1850 CZK
ACRO463020010 ACRO463020050
(+)-Catechin monohydrate
(+)-Catechin monohydrate

  • Bioactive

Vzorec: C₁₅H₁₄O₆·H₂O
MW: 290.3 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00075649
CAS číslo: 225937-10-0

Objednat

Výsledky specifikačního testu

Appearance White to beige to light brown Powder
Infrared spectrum Conforms
Loss on drying ≤8 %
Solubility (5 % in Ethanol, hot) Clear to hazy
HPLC ≥96.0 %
Specific optical rotation +24° to +28° (20°C, 589 nm) (1 % in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR