Tisk…

Nateglinide

Dodavatel: Thermo Fisher Scientific

Synonyms: N-[(trans-4-Isopropylcyclohexyl)carbonyl]-D-phenylalanine, (R)-2-rel-((1r,4R)-4-Isopropylcyclohexanecarboxamido)-3-phenylpropanoic acid

462980010 462980050
ACRO462980010EA 2490 CZK
ACRO462980010 ACRO462980050
Nateglinide
Nateglinide

  • Bioactive

Vzorec: C₁₉H₂₇NO₃
MW: 317.43 g/mol
Bod tání: 129…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00875706
CAS číslo: 105816-04-4

Objednat

Výsledky specifikačního testu

Appearance White to almost white Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (1 g, 105°C, 2 h)
HPLC ≥97.5 %
Specific optical rotation -36.5° to -40° (20°C, 589 nm) (1 % in 0.1M NaOH)
Sulfate (SO4) ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR