Tisk…

Labetalol hydrochloride ≥99%

Dodavatel: Thermo Fisher Scientific

462600050 462600250
ACRO462600050EA 3940 CZK
ACRO462600050 ACRO462600250
Labetalol hydrochloride ≥99%
Labetalol hydrochloride

  • Bioactive

Vzorec: C₁₉H₂₄N₂O₃·HCl
Storage Temperature: Ambient
MDL Number: MFCD00057663
CAS číslo: 32780-64-6

Objednat

Výsledky specifikačního testu

Appearance White to almost white Powder
Infrared spectrum Conforms
Melting point 178 to 180 °C
Loss on drying ≤1 %
Solubility (250 mg + 5 ml methanol) Clear to light hazy colorless to light yellow-pink solution
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR