Tisk…

Fenbendazole

Dodavatel: Thermo Scientific

Synonyms: Methyl [5-(Phenylthio)benzimidazol-2-yl]carbamate, methyl (6-(phenylthio)-1H-benzo[d]imidazol-2-yl)carbamate, [5-(Phenylthio)benzimidazol-2-yl]carbamic Acid Methyl Ester

462470250 462471000
ACRO462470250EA 3210 CZK
ACRO462470250 ACRO462471000
Fenbendazole
Fenbendazole

  • Bioactive

Vzorec: C₁₅H₁₃N₃O₂S
MW: 299,35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00144301
CAS číslo: 43210-67-9

Objednat

Výsledky specifikačního testu

Appearance White to beige Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR