Tisk…

Esmolol hydrochloride

Dodavatel: Thermo Scientific
Varování

Synonyms: 3-[4-[2-Hydroxy-3-(isopropylamino)propoxy]phenyl]propionic acid methyl ester hydrochloride, Methyl 3-[4-[2-Hydroxy-3-(isopropylamino)propoxy]phenyl]propionate hydrochloride

462460050 462460250
ACRO462460050EA 2480 CZK
ACRO462460050 ACRO462460250
Esmolol hydrochloride
Esmolol hydrochloride

  • Bioactive

Vzorec: C₁₆H₂₆ClNO₄
MW: 331,84 g/mol
Bod tání: 86…92 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00941432
CAS číslo: 81147-92-4

Objednat

Výsledky specifikačního testu

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 86 to 92 °C
Water ≤1 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR