Tisk…
Ziprasidone hydrochloride monohydrate
Dodavatel: Thermo Fisher Scientific
Synonyms: 5-[2-[4-(1,2-Benzisothiazol-3-yl)-1-piperazinyl]ethyl]-6-chloro-1,3-dihydro-2H-indol-2-one Hydrochloride
462430010 462432500
ACRO462430010EA 18530 CZK
ACRO462430010 ACRO462432500
Ziprasidone hydrochloride monohydrate
Ziprasidone hydrochloride monohydrate
  • Bioactive
Vzorec: C₂₁H₂₁ClN₄OS·HCl·H₂O
MW: 449.39 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00921885
CAS číslo: 138982-67-9
Objednat
Výsledky specifikačního testu
Appearance White to light pink or light orange Powder
Infrared spectrum Conforms
Water 3.7 to 5.0 %
HPLC ≥97.5 %