Sumatriptan succinate
Dodavatel: Thermo Scientific
|
Nebezpečí
|
462420010
462420050
ACRO462420010EA
6810
CZK
ACRO462420010
ACRO462420050
Sumatriptan succinate
Sumatriptan succinate
- Bioactive
Vzorec:
C₁₄H₂₁N₃O₂S Bod tání: 165…166 °C Storage Temperature: Ambient |
MDL Number:
MFCD00902856 CAS číslo: 103628-48-4 |
Výsledky specifikačního testu
Appearance | White to almost white Powder |
Infrared spectrum | Conforms |
Melting point | 165 to 169 °C |
Water | ≤1 % |
Solubility | (2.5 % in water) Clear colorless solution |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...