Tisk…

Propafenone hydrochloride

Dodavatel: Thermo Scientific
Varování

Synonyms: 1-(2-(2-Hydroxy-3-(propylamino)propoxy)phenyl)-3-phenylpropan-1-one hydrochloride, Propafenone HCl

462390010 462390050
ACRO462390010EA 2090 CZK
ACRO462390010 ACRO462390050
Propafenone hydrochloride
Propafenone hydrochloride

  • Bioactive

Vzorec: C₂₁H₂₇NO₃·HCl
MW: 377,9 g/mol
Bod tání: 173 °C
Storage Temperature: Ambient
MDL Number: MFCD00079243
CAS číslo: 34183-22-7

Objednat

Výsledky specifikačního testu

Appearance Colorless to white Powder
Infrared spectrum Conforms
Melting point 171 to 175 °C
Loss on drying ≤0.5 %
Solubility (5 % in methanol) Clear colorless solution
pH 5.0 to 6.2 (0.5 % in water)
Titration with HClO4 98 to 102 % (on dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR