Tisk…

Vinorelbine tartrate ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Methyl (3aR,3a1R,4R,5S,5aR,10bR)-4-acetoxy-3a-ethyl-9-((8R)-4-ethyl-8-(methoxycarbonyl)-1,3,6,7,8,9-hexahydro-2,6-methanoazecino[4,3-b]indol-8-yl)-5-hydroxy-8-methoxy-6-methyl-3a,3a1,4,5,5a,6,11,12-octahydro-1H-indolizino[8,1-cd]carbazole-5-carboxylate bis((2R,3R)-2,3-dihydroxysuccinate), Vinorelbine ditartrate

461980250 461981000
ACRO461980250EA 5080 CZK
ACRO461980250 ACRO461981000
Vinorelbine tartrate ≥98%
Vinorelbine tartrate

  • Bioactive

Vzorec: C₅₃H₆₆N₄O₂₀
MW: 1079.106 g/mol
Storage Temperature: Freezer
MDL Number: MFCD03613607
CAS číslo: 125317-39-7
UN: 2811
ADR: 6.1 III

Objednat

Výsledky specifikačního testu

Appearance White to almost white Powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR