Asiaticoside ≥92%
Dodavatel: Thermo Scientific
Synonyms:
(1S,2R,4aS,6aS,6bR,8aR,9R,10R,11R,12aR,12bR,14bS)-(2S,3R,4S,5S,6R)-6-((((2R,3R,4R,5S,6R)-3,4-Dihydroxy-6-(hydroxymethyl)-5-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)methyl)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl 10,11-dihydroxy-9-(hydroxymethyl)-1,2,6a,6b,9,12a-hexamethyl-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-icosahydropicene-4a-carboxylate
- Bioactive
Vzorec:
C₄₈H₇₈O₁₉ MW: 959,12 g/mol Bod tání: 230…233 °C Storage Temperature: Ambient |
MDL Number:
MFCD06642601 CAS číslo: 16830-15-2 |
Výsledky specifikačního testu
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 228 to 234 °C |
Loss on drying | ≤2 % |
HPLC | ≥92.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...