Tisk…

Mitotane

Dodavatel: Thermo Scientific

Synonyms: o,p'-DDD, 2,4'-TDE, o,p'-TDE, 2,4'-DDD

109850010 109850050
ACRO109850010EA 1440 CZK
ACRO109850010 ACRO109850050
Mitotane
Mitotane

  • Bioactive

Vzorec: C₁₄H₁₀Cl₄
MW: 320,04 g/mol
Bod tání: 77…78 °C
Storage Temperature: Freezer
MDL Number: MFCD00000850
CAS číslo: 53-19-0

Objednat

Výsledky specifikačního testu

Appearance White to beige Powder or crystals
Infrared spectrum Conforms
Melting point 74 to 80 °C
Loss on drying ≤1 %
HPLC ≥96.0 %
Silicon dioxide (SiO2) ≤0.03 %
Iron(III)oxide (Fe2O3) ≤0.03 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR