Tisk…

Phenol red sodium salt, AnalaR NORMAPUR® ACS indicator

Dodavatel: VWR Chemicals

85920.130EA 3070 CZK
85920.130 85920.100
Phenol red sodium salt, AnalaR NORMAPUR® ACS indicator
Phenol red sodium salt
Vzorec: C₁₉H₁₃NaO₅S
MDL Number: MFCD00066901
CAS číslo: 34487-61-1

Objednat

Výsledky specifikačního testu

Appearance Dark red to brown powder
Visual transition interval pH 6.5 - pH 8.2
Loss on drying (110°C) ≤ 5 %
Absorption max. λ 1 (pH 6.5) 430 - 435 nm
Absorption max. λ 2 (pH 8.2) 557 - 560 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR