Tisk…
Carbolic auramine
Dodavatel: RAL (REACTIFS RAL SA)
720-0345EA 2070 CZK
720-0345
Carbolic auramine
Carbolic auramine
RAL Auramine staining kit allows fluorescence detection of Acid Fast Bacilli (AFB). The A.F.B appear fluorescent-yellow on the blue background preparation.
Used in Auramine staining kit Ref 359500
IVDLogo