Tisk…
Nitrile gloves, powder-free, Viva®
Dodavatel: SRI TRANG GLOVES
7024770 7027749 7027729 7027731 7027732
SRIT7024770EA 827 CZK
SRIT7024770 SRIT7027749 SRIT7027729 SRIT7027731 SRIT7027732
Nitrile gloves, powder-free, Viva®
Rukavice
High-quality, dual labelled PPE and MDR nitrile gloves.
  • Non sterile, powder-free
  • Low thickness and textured fingertips for excellent grip
  • Particularly skin friendly for users with latex allergy
  • Rolled cuffs minimise the risk of tearing
  • Free from phthalates, softeners, and allergenic latex proteins
These light, ambidextrous, durable gloves are ideal for medical and lab applications, cleaning, and other tasks with low risks
Certifikace: EN 455, EN ISO 374-1, EN 374-2, EN 374-4, EN 374-5, EN 16523-1; Cat. III; AQL 1,5
Food contact approved
Objednat
Produktová specifikace
Obouruké/jednoruké Obouruké
Environmentally Preferable
Jednorázové/znovupoužitelné Jednorázové
Latex/bez latexu Latex-free
Pudrované/nepudrované Nepudrované
Materiál Nitril
Provedení Textured fingertips
Typ manžety Rolled
Tloušťka prstu 0,05 mm
Tloušťka drapáku 0,05 mm
Tažná síla 14 MPa
Konečné prodloužení 400 %
Norma EN EN 455, EN ISO 374-1, EN 374-2, EN 374-4, EN 374-5, EN 16523-1
Schváleno pro styk s potravinami +