Tisk…
Cleanroom gloves, polychloroprene, sterile, BioClean Ultimate™
Dodavatel: Nitritex
BUPS-80 BUPS-75 BUPS-90 BUPS-60 BUPS-65 BUPS-70 BUPS-85
112-1664EA 17490 CZK
112-1664 112-1663 112-1666 112-1660 112-1661 112-1662 112-1665
Cleanroom gloves, polychloroprene, sterile, BioClean Ultimate™
Rukavice
BioClean Ultimate™ sterile cleanroom gloves are manufactured from polychloroprene and are hand-specific with a beaded cuff. They offer excellent cytotoxic protection with sensitivity.
  • Sterile, powder-free, textured finish provides a good grip
  • With no latex proteins, Type 1 latex allergic reactions are eliminated
  • Anatomical hand-specific shape for high tactility, protection and comfort
  • Fit and feel of natural latex gloves
Suitable for use in Class 10, ISO 4 or M2.5 and EU GMP Grade A cleanrooms.
Colour: Natural
EN 420, EN 455, Cat III, AQL: 1.5
Certifikace: Manufactured in a facility holding ISO 9001:2015 and (EU)2016/425 Module D certifications.
Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Regulation (EU)2016/425.
Processed in an NEBB certified ISO Class 4 environment.
Complies with the requirements of European Glove Standards EN 420:2003 + A1:2009, EN 374-1:2016, EN 374-5:2016 and EN 421:2010.
Physical properties comply with European Medical Glove Standard EN 455-2:2015.
Sterilised in accordance with ISO 11137-1:2006.
Suitable for contact with all foodstuffs. Regulation (EC)1935/2004 and (EC) 2023/2006.
Tested against permeation standard ASTM D6978-05 for cytotoxic drugs.
Balení: One cuffed pair per inner plastic wallet; one inner wallet per EasyTear™ PE pouch; 10 pairs per sealed outer PE bag; 20 outer bags per carton liner; one carton liner per carton (200 pairs).