Tisk…
Visual comparator test kits, dissolved oxygen by rhodazine method, CHEMets
Dodavatel: CHEMetrics
K-7540 R-7540
CHETK-7540EA 9390 CZK
CHETK-7540 CHETR-7540
Visual comparator test kits, dissolved oxygen by rhodazine method, CHEMets
Kity pro environmentální testy
Test kits for boiler waters and applications requiring trace levels of D.O.
  • Rhodazine D method
  • Salinity, dissolved gas, hydrazine and sulfite do not interfere
Informace o dodávce: Kit comes in a plastic case and contains everything needed to perform 30 tests: Refill, Comparator, adhesive mounting clamp, permanent mounting clamp, sampling tube, instructions, and MSDS.