Tisk…
Safety ankle boots, lace-up, Compositelite™ Thor, FC11
Dodavatel: Portwest
FC11BKR39 FC11BKR45 FC11BKR46 FC11BKR41 FC11BKR38 FC11BKR43 FC11BKR44 FC11BKR47 FC11BKR48 FC11BKR40 FC11BKR42
PRTWFC11BKR39EA 2330 CZK
PRTWFC11BKR39 PRTWFC11BKR45 PRTWFC11BKR46 PRTWFC11BKR41 PRTWFC11BKR38 PRTWFC11BKR43 PRTWFC11BKR44 PRTWFC11BKR47 PRTWFC11BKR48 PRTWFC11BKR40 PRTWFC11BKR42
Safety ankle boots, lace-up, Compositelite™ Thor, FC11
Obuv
Metal-free, anti-static, lightweight boot made from water resistant leather fabric plus a durable, dual-density PU/rubber sole, resistant to oil and heat (300 °C).
  • Shock-absorbent heel
  • Composite toecap (200 J)
  • Puncture resistant composite midsole
Top class design and engineering that offers stylish and durable protection. Non metallic safety components ideal for industrial wear with a strong and lightweight construction.
Colour: Black
Certifikace: EN ISO 20345; S3; SRC certified.