Tisk…
Cut resistant gloves, A723
Dodavatel: Portwest
A723G8RL A723G8RM A723G8RXL A723G8RXXL
PRTWA723G8RLEA 6680 CZK
PRTWA723G8RL PRTWA723G8RM PRTWA723G8RXL PRTWA723G8RXXL
Cut resistant gloves, A723
Rukavice
This cut C glove has an added layer of protection, using an innovative TPV back protection. The nitrile micro foam palm gives superior grip and protection against liquids and water penetration.
  • 13 gauge liner for a perfect fit
  • Reinforced padded palms
  • Level C cut resistance
  • 100% breathable seamless liner
  • Impact resistance and high flexibility
Low linting construction for minimal contamination. Ergonomic design for optimised comfort.
Materials: TPV, HDPE, nitrile foam.
Certifikace: Standards: AS/NZS 2161 .3, AS/NZS 2161 .2, EN 420 (Dexterity 4), EN388:2016 +A1:2018 - (4.X.4.3.C.P.)