Germanium ICP Standards, (CRMs), Fluka™
Dodavatel: Honeywell Chemicals
E1024-100ML
E1151-100ML
HONCE1024-100MLEA
2910
CZK
HONCE1024-100ML
HONCE1151-100ML
Germanium ICP Standards, (CRMs), Fluka™
Standardy
Standardy kationické
Standardy - germanium
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Certifikace: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).